NanoDrop™ PR-1 Reconditioning Kit - FAQs
NanoDrop™ PR-1 Reconditioning Kit - FAQs
View additional product information for NanoDrop™ Reagents - FAQs (CHEM-PV-8, CHEM-CF-8, CHEM-PR1-KIT, CHEM-PV-1, CHEM-CF-1)
3 product FAQs found
How do I keep my sample from spreading on the pedestal of my NanoDrop Lite Spectrophotometer?
There are many reagents and organic solvents that could compromise the pedestal surface properties, causing samples to flatten out rather than bead up. Use the NanoDrop PR-1 reconditioning compound as a rapid means of reconditioning the pedestals when the pedestal surface properties have been compromised and samples spread out on the pedestal. NanoDrop PR-1 Reconditioning Kits are available through Thermo Fisher website (Cat. No. CHEM-PR1-KIT)
My NanoDrop 8000 Spectrophotometer failed calibration check. What should I do?
Please review the following steps to take when/if your NanoDrop 8000 Spectrophotometer fails a calibration check:
- Clean the pedestal with the NanoDrop PR-1 Reconditioning Kit
- Repeat calibration check using a FRESH vial of NanoDrop CF-1 Calibration Fluid and FRESH blanking solution
- If instrument continues to be out of specification, Contact Technical Support
My NanoDrop 2000/2000c Spectrophotometer failed the calibration check. What should I do?
Please review the following steps to take when/if your NanoDrop 2000/2000c fails a calibration check:
- Clean the pedestal with the NanoDrop PR-1 Reconditioning Kit
- Repeat calibration check using a FRESH vial of NanoDrop CF-1 Calibration Fluid and FRESH blanking solution
More detailed instructions can be found on the Calibration Check Procedure document: https://assets.thermofisher.com/TFS-Assets/CAD/Product-Bulletins/NanoDrop-2000-2000c-1000-Calibration-Check-procedure.pdf
If your NanoDrop 2000/2000c continues to be out of specification, Contact Technical Support.