Arctic Express™ Dual Storage Systems
Actual product may vary
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Thermo Scientific™

Arctic Express™ Dual Storage Systems

Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CK5092228 L
CK5092010 L
CK5092118.5 L
Catalog number CK50922
Price (MXN)
-
Capacity (Metric):
28 L
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
  • Warranty varies by country, please contact us for details
Specifications
Diameter (English)18.2 in.
Diameter (English) Inside Neck3.8 in.
Diameter (Metric)46.2 cm
Diameter (Metric) Inside Neck9.7 cm
ModelArctic Express Dual 28
Static Holding Time21 shipper/50 lab
TypeManual Fill LN2 Dewar
Vial Capacity630 x 2mL
Capacity (Metric)28 L
No. of Doors1
Unit SizeEach