CleanMill Broad Ion Beam System
Thermo Scientific™
CleanMill Broad Ion Beam System
The CleanMill Broad Ion Beam System (cross section polisher) offers ion beam polishing and ion beam milling of air-sensitive and other samples for SEM imaging.
製品番号(カタログ番号)タイプ
CLEANMILLFIB-SEM
製品番号(カタログ番号) CLEANMILL
価格(JPY)
-
タイプ:
FIB-SEM
High-quality observation and characterization of materials often require an artifact-free surface, which can be difficult to achieve with traditional polishing techniques like grinding or mechanical polishing.
The Thermo Scientific™ CleanMill Broad Ion Beam System is a complete ion beam polishing solution for SEM applications in materials science, enabling optimal imaging and analysis of materials where a pristine surface is required, including beam- and air-sensitive materials.
The CleanMill System is fully compatible with the Thermo Scientific™ CleanConnect Sample Transfer System, making it easy to quickly transfer samples between instruments while minimizing sample handling.
Find more information: CleanMill Broad Ion Beam System
CleanMill Broad Ion Beam System features
High-energy ion source
The ultra-high-energy ion gun features a maximum accelerating voltage of 16 kV to rapidly mill and polish sample surfaces.
Ultra-fine surface polishing
The CleanMill System can be configured with an optional low-energy io n gun for final polishing of sample surfaces.
Wide acceleration voltage
The system delivers ion energy ranging from 2 kV to 16 kV and features dedicated optics for ultra-low-voltage polishing from 100 V to 2 kV.
CleanConnect System compatibility
Safely transfer samples from the CleanMill System to the microscope chamber using the Thermo Scientific IGST (inert gas sample transfer) Workflow to observe materials in their native states.
Real-time monitoring
The integrated touchscreen and high-resolution camera help you keep track of the broad ion beam milling process.
Cryogenic milling
The optional cryo-stage delivers LN2 cooling with automatic refilling for working with extremely beam-sensitive materials.
仕様
概要CleanMill Broad Ion Beam System (cross section polisher) Ion beam polishing and ion beam milling for SEM imaging
タイプFIB-SEM
Unit SizeEach