Nutrient Broth (Dehydrated)
Thermo Scientific™
Nutrient Broth (Dehydrated)
Cultivate fastidious pathogens and other microorganisms with this nutritious medium.
製品番号(カタログ番号)数量
CM0001B500 g
CM0001R2.5 kg
製品番号(カタログ番号) CM0001B
価格(JPY)
-
数量:
500 g
Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth (Dehydrated). This general purpose medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
A general purpose fluid medium for the cultivation of microorganisms not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
仕様
概要Nutrient Broth
数量500 g
収量For 38.5L medium
形状粉末
製品タイプDehydrated Culture Media
Unit SizeEach