This product is no longer available to order. Alternative product CM0681B or CM0681R is available. Click the link to find out more.
Milk Agar (Dehydrated)
Thermo Scientific™
Milk Agar (Dehydrated)
Determine the viable microflora of dairy and water samples with this medium corresponding to the medium described in Dept. Of Health Memo.139/Foods.
Have Questions?
Catalog NumberQuantity
CM0021B500 g
Catalog number CM0021B
Price (KRW)
-
Quantity:
500 g
Determine the viable microflora of dairy and water samples with Thermo Scientific™ Oxoid™ Milk Agar (Dehydrated) – a nutrient medium enriched with milk solids. Milk Agar is made to a formula corresponding to the official medium described in Dept. of Health Memo. 139/ Foods1. It is recommended for performing the plate count test on milks, rinse waters, milk products and ice cream, etc.
It also complies with the recommendations of EUROGLACE (EEC Ice cream Industries) submitted to the EEC Commission for the examination of ice cream2.
Liquid milk and other dairy products are highly perishable and can spoil in a few days. The bacteria present in dairy products may cause disease or spoilage. Foodborne illness from dairy product consumption of have been predominantly associated with Salmonella enterica, Listeria monocytogenes, Camplylobacter jejuni and Escherichia coli O157: H7.
Milk Agar is a nutrient medium enriched with milk solids for the determination of the viable micro-flora of dairy and water samples.
  • Corresponds to the official medium described in Dept. Of Health Memo.139/Foods1
  • Recommended for performing the plate count test on milks, rinse waters, milk products and ice cream, etc.
  • Complies with the recommendations of EUROGLACE (EEC Ice cream Industries) submitted to the EEC Commission for the examination of ice cream2
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
General References:
  1. Dept. of Health (1987) Memo. 139/Foods.
  2. Klose J. (1968) Susswaren. 14. 778-782.
Specifications
DescriptionMilk Agar
Quantity500 g
YieldFor 20.8L medium
FormPowder
Product TypeAgar
Unit SizeEach