Urea Agar Base (Dehydrated)
Thermo Scientific™

Urea Agar Base (Dehydrated)

Detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae with this medium.

Have Questions?
카탈로그 번호수량
CM0053B500 g
카탈로그 번호 CM0053B
제품 가격(KRW)
-
수량:
500 g

Detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae with Thermo Scientific™ Oxoid™ Urea Agar Base (Dehydrated).

Urea Agar Base is recommended for the preparation of Christensen medium1 for the detection of rapid urease activity of the urease-positive Proteae. The urea medium may be used for the detection of urea hydrolysis by some other Enterobacteriaceae but the incubation period is much longer 24-48 hours.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

For Laboratory Use Only
사양
인증/적합성Industrial Reference: AOAC, FDA.
설명Urea Agar Base
수량500 g
수율For 20.8L medium
형태Powder
제품 유형Agar
Unit SizeEach