Oxoid™ Blood Agar Base (Dehydrated)
Thermo Scientific™

Oxoid™ Blood Agar Base (Dehydrated)

Grow a wide variety of pathogenic and non-pathogenic bacteria with this highly nutritious general purpose medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0055T5 kg
CM0055B500 g
Catalog number CM0055T
Price (KRW)
-
Quantity:
5 kg

Grow a wide variety of pathogenic and non-pathogenic bacteria with general purpose Thermo Scientific™ Oxoid™ Blood Agar Base (Dehydrated). Without any additions, the medium can be used for short-term maintenance of stock cultures. With added serum and other enrichments, the medium is suitable for cultivation of many fastidious organisms. With the addition of blood, the medium becomes suitable for the determination of hemolytic reactions which are important diagnostic criteria for many organisms.

Blood Agar Base is a highly nutritious medium generally used as a basal medium for preparing blood agar by the addition of blood. It can also be used as general purpose medium without the addition of blood.

Blood Agar Base was used during investigations on irradiated Escherichia coli and other bacteria1,2. It was the most suitable medium for investigating the phages of Clostridium perfringens3 and as the basis of a selective medium for Clostridium perfringens4. It was used with added phenolphthalein phosphate for the detection of phosphatase-producing staphylococci5 and with added salt and agar for the assessment of surface contamination on equipment and pig carcasses6. It has been used for determining the salinity range of growth of marine flavobacteria7.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For Laboratory Use Only

General References:

  1. Alper T. and Gillies N. E. (1960) J. Gen. Microbiol. 22 113-128.
  2. Hodgkins Brenda and Alper T. (1963) J. Gen. Microbiol. 30 307-315.
  3. Williams Smith H. (1959) J. Gen. Microbiol. 21 622-630.
  4. Noble W. C. (1961) J. Path. Bact. 81 523-526.
  5. Noble W. C. (1962) J. Clin. Path. 15 552-558.
  6. Hansen N. H. (1962) J. Appl. Bact. 25 46-53.
  7. Hayes P. R. (1963) J. Gen. Microbiol. 30 1-19
Specifications
Quantity5 kg
FormPowder
Product TypeAgar
Unit SizeEach