Tryptone Soya Agar EP/USP/JP/BP (Dehydrated)
Thermo Scientific™
Tryptone Soya Agar EP/USP/JP/BP (Dehydrated)
Grow a wide variety of microorganisms with this general purpose medium specified in international pharmacopoeia.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
CM0131K25 kg
CM0131T5 kg
CM0131B500 g
CM0131R2.5 kg
Catalog number CM0131K
Price (USD)
-
Quantity:
25 kg
Grow a wide variety of microorganisms with Thermo Scientific ™ Oxoid ™ Tryptone Soya Agar (Dehydrated). A general purpose agar medium containing two peptones, Tryptone Soya Agar supports the growth of both aerobes and anaerobes, the latter being grown either in deep cultures or by incubation under anaerobic conditions. Tryptone Soya Agar is recommended as a reference medium when testing selective media to measure the degree of inhibition1.
Tryptone Soya Agar, also known as Casein Soya Bean Digest Agar, is a general purpose medium specified in pharmacopoeia and food testing methods for the growth of a wide variety of organisms. The product is tested to the pharmacopoeia2, 3, 4, 5 and is mainly used as reference medium when testing selective media.
  • Recommended as a reference medium for testing selective media
  • Conforms to formulations detailed in the International pharmacopoeia
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use Only
1. Anon. (1987) J. Food Microbiol. 5. 291-296.
2. British Pharmacopoeia Volume II (2000)
3. US Pharmacopoeia XXX, (2008)
4. European Pharmacopoeia. 6.1 Edition (2008)
5. Japanese Pharmacopoeia. 15th Edition. (2006)
Specifications
Quantity25 kg
FormPowder
Product TypeTryptone Soya Agar
Unit SizeEach