D.C.L.S. Agar (Dehydrated)
Thermo Scientific™
D.C.L.S. Agar (Dehydrated)
Isolate and enumerate pathogenic Enterobacteriaceae with this selective medium.
Have Questions?
Catalog number CM0393B
Price (USD)
-
Isolate and enumerate pathogenic Enterobacteriaceae with Thermo Scientific™ Oxoid™ D.C.L.S. Agar (Dehydrated). Due to the presence of sucrose in the medium, sucrose fermenting organisms such as Proteus, Enterobacter and Klebsiella form red colored colonies. Due to the selective nature of the medium, it allows the growth of Vibrio species, as well as salmonellae and shigellae, while inhibiting the growth of most Escherichia coli.
Use D.C.L.S. Agar for improved identification of pathogenic Enterobacteriaceae.
  • Easy-to-read: sucrose fermenting organisms such as Proteus, Enterobacter and Klebsiella form red colored colonies
  • Reduces false-positive subcultures when picking through color recognition.
  • Selects for salmonellae and shigellae, while inhibiting Escherichia coli.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Certifications/ComplianceIndustrial Reference: AFNOR, ISO, NMKL.
DescriptionD.C.L.S. Agar
Quantity500 g
YieldFor 10.0L medium
FormPowder
Product TypeAgar
Unit SizeEach