Clostridium Difficile Agar Base (Dehydrated)
Thermo Scientific™

Clostridium Difficile Agar Base (Dehydrated)

Isolate Clostridium difficile from fecal specimens with this medium when used with Culture Media Supplement (SR0096).

Have Questions?
카탈로그 번호수량
CM0601B500 g
카탈로그 번호 CM0601B
제품 가격(KRW)
-
수량:
500 g

Isolate Clostridium difficile from fecal specimens with Thermo Scientific™ Oxoid™ Clostridium Difficile Agar Base (Dehydrated) when used with Culture Media Supplement (SR0096). The combination of Clostridium Difficile Agar Base and the Culture Media Supplement (SR0096) is based on the formulation proposed by George et al.1 The selective agents D-cycloserine (500μg/ml) and cefoxitin (16μg/ml) inhibit growth of the majority of non target organisms which may be found in large numbers in fecal samples.

Clostridium difficile was first isolated in 1935 by Hall and O’Toole2 who proposed the name `difficile’ because it was very difficult to isolate. Clostridium difficile is a toxin-producing, spore forming anaerobic gram-positive rod, which causes gastrointestinal infections ranging in severity from asymptomatic colonization to severe diarrhea, pseudomembranous colitus (PMC), toxic megacolon and colonic perforaton.

Clostridium Difficile Agar Base is used for the primary isolation of C. difficile from fecal specimens. The medium composition is designed so as to obtain luxuriant growth of C. difficile.

  • Selective: the selective agents D-cycloserine (500μg/mL) and cefoxitin (16μg/mL) inhibit growth of the majority of Enterobacteriaceae, as well as Streptococcus fecalis, staphylococci, Gram-negative non-sporing anaerobic bacilli and Clostridia (except Clostridium difficile) which may be found in large numbers in fecal samples.
  • Easy-to-read: C. difficile forms grey-white colored colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

체외진단 전용.
For In Vitro Diagnostic Use.  

General References:

  1. George W. L., Sutter V. L., Citron D. and Finegold S. M. (1979) J. Clin. Microbiol. 9. 214-219.
  2. Hall I. and O'Toole E. (1935) Am. J. Dis. Child. 49. 390.
사양
설명Clostridium difficile Agar Base
수량500 g
수율For 7.2L medium
형태Powder
제품 유형Agar
Unit SizeEach