Minerals Modified Glutamate Medium (Dehydrated)
Minerals Modified Glutamate Medium (Dehydrated)
Minerals Modified Glutamate Medium (Dehydrated)
Thermo Scientific™
Minerals Modified Glutamate Medium (Dehydrated)
Identify members of the coliform group of bacteria in food and water samples with this medium.
Have Questions?
Número do catálogoFormProduct Type
CM0607GPowderDehydrated Culture Media
Número do catálogo CM0607G
Preço (BRL)
-
Form:
Powder
Product Type:
Dehydrated Culture Media
Identify members of the coliform group of bacteria in food and water samples with Thermo Scientific™ Oxoid™ Minerals Modified Glutamate Medium (Dehydrated). The mineral content of the medium allows for the superior cultivation and enumeration of coliforms.
Use Minerals Modified Glutamate Medium to identify coliforms in food and water samples by the technique known as the Multiple Tube Method, Dilution Method, or the Most Probable Number (MPN) method.
  • Superior performance over MacConkey Broth in the detection of Escherichia coli.
  • Medium of choice for detection of small numbers of Escherichia coli in chlorinated water.
  • Effectively isolates Escherichia coli from sea water.
Minerals Modified Glutamate Medium was developed by Oxoid Laboratories and Metropolitan Water Board Laboratories and based upon the formulation described by Gray1
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
1. Gray R. D. (1959) J. Hyg. Camb. 57. 249-265.
Especificações
QuantityEach
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Conteúdo e armazenamento
10°C to 30°C