Maximum Recovery Diluent (Dehydrated)
Thermo Scientific™
Maximum Recovery Diluent (Dehydrated)
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic diluent.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
CM0733R2,5 kg
CM0733B500 g
Número de catálogo CM0733R
Precio (CLP)
-
Cantidad:
2,5 kg
Achieve maximum recovery of microorganisms during dilution with this protective, isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent (Dehydrated).
Maximum Recovery Diluent combines the protective effect of peptone in the diluting solution1 with the osmotic support of physiological saline1,2. The low peptone concentration in the diluent reduces organism multiplication in the sample for at least 45 minutes during the dilution stage1,2
Maintain the viability of organisms during dilution process with Maximum Recovery Diluent with low levels of peptone.
  • Allows accurate quantitative procedures to be performed with minimal reduction in viable count.
  • Reduces organism multiplication in the sample for at least one hour during the dilution stage.
The presence of peptone in the diluent provides viable counts to ensure accurate quantitative procedures are performed. The isotonic strength of the diluent ensures recovery of organisms from various sources which may be vulnerable in distilled water or aqueous suspensions.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Especificaciones
Certificaciones/conformidadIndustrial Reference: AFNOR, BSI, EP, IDF, ISO, NMKL
Cantidad2,5 kg
FormularioPowder
Tipo de productoDehydrated Culture Media
Unit SizeEach