Oxoid™ Listeria Enrichment Broth
Thermo Scientific™

Oxoid™ Listeria Enrichment Broth

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.
Have Questions?
카탈로그 번호수량
CM0862B500 g
카탈로그 번호 CM0862B
제품 가격(KRW)
-
수량:
500 g

Oxoid Listeria Enrichment Broth is used for the enrichment of Listeria species.

Also available Listeria Selective Supplement, Part No. SR0140E or SR0141E and Modified Listeria Selective Enrichment Supplement, Part No. SR0206E or SR0206B.
For Laboratory Use Only
사양
인증/적합성Industrial Reference: FDA, IDF.
설명Listeria Enrichment Broth
수량500 g
수율For 13.9L medium
형태Powder
제품 유형Dehydrated Culture Media
Unit SizeEach