Search
Search

Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014
| Número de catálogo | Cantidad |
|---|---|
| CM1049T | 5 kg |
| CM1049B | 500 g |
| CM1049G | 10 kg |
| CM1049K | 25 kg |
| CM1049R | Cada uno |
Pre-enrich food samples prior to the isolation of Salmonella species using Thermo Scientific™ Oxoid™ Buffered Peptone Water (ISO) (Dehydrated). Buffered Peptone Water is a non-selective pre-enrichment medium and is tested in accordance with ISO 65791 and ISO 11133:20142. It also provides conditions for resuscitation of cells that have been injured by processes of food preservation.
Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.