Chromogenic Coliform Agar (Dehydrated)
Chromogenic Coliform Agar (Dehydrated)
Chromogenic Coliform Agar (Dehydrated)
Thermo Scientific™

Chromogenic Coliform Agar (Dehydrated)

Detect, enumerate and differentiate coliforms and E.coli in water samples using Chromogenic Coliform Agar in accordance with ISO 9308-1.
Have Questions?
Catalog NumberFormQuantity
CM1205BPowder500 g
Catalog number CM1205B
Price (CLP)
-
Form:
Powder
Quantity:
500 g
Enumerate and differentiate coliforms and Escherichia coli in water samples with Thermo Scientific™ Oxoid™ Chromogenic Coliform Agar in accordance with ISO 9308-1 guidelines for water quality analysis. Target organisms are easy to differentiate, E.coli forming dark blue/violet colonies and coliforms pink to red colonies. Achieve complete detection within 24 hours, removing the need for subculturing.

Chromogenic Coliform Agar is a fast, accurate, and efficient way to detect coliforms and E.coli during microbiological quality testing of drinking water, disinfected pool water, and treatment plant samples. The guideline, ISO 9308-1:2014 Enumeration of Escherichia coli and coliform bacteria –Part 1: Membrane filtration method for waters with low bacterial background flora, specifies a method based on membrane filtration, subsequent culture on a Chromogenic Coliform Agar medium, and calculation of the number of target organisms in the sample.

Chromogenic Coliform Agar utilizes specific enzymes associated with general coliforms and E.coli for differentiation.

  • The chromogen Salmon GAL, when cleaved by D-galactosidase, produces a pink to red coloration; this coloration and enzyme are associated with coliforms.
  • E.coli contains D-galactosidase, which cleaves Salmon GAL; however, it also contains D-glucuronidase, which cleaves the chromogen X-glucuronide. The combination of these chromogens produces a dark blue to violet coloration associated with E.coli.
  • Tergitol-7 within the medium inhibits Gram positive bacteria and Gram negative bacteria with exception of coliforms.
For Laboratory Use
For Laboratory Use
Specifications
DescriptionChromogenic Coliform Agar (Dehydrated)
Quantity500 g
Shelf Life3 Years
FormPowder
Product TypeAgar
Unit SizeEach
Contents & Storage
10°C to 30°C