IL-12 p70 Mouse Matched Antibody Pair

Catalog number: CMC0123

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of mouse IL-12. Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Buffer reagents needed to complete the reaction are sold separately under Catalog #CNB0011.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: IL-12p35, Il-12a, Ll12a, p35, Il-12b, Il-12p40, Il12p40, p40
Gene ID: 16159, 16160
Gene Name: interleukin 12A, interleukin 12b
Gene Symbol: IL12A, IL12B
Immunoassay Kit Format: Matched Antibody Pair
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 10 x 96 tests
Protein Aliases: IL-12A, Cytotoxic lymphocyte maturation factor 35 kDa subunit, CLMF p35, IL-12 subunit p35, IL-12B, Cytotoxic lymphocyte maturation factor 40 kDa subunit, CLMF p40, IL-12 subunit p40
Protein Name: Interleukin-12 subunit alpha, Interleukin-12 subunit beta
Shipping Condition: Approved for shipment on Wet or Dry Ice
Target (Kit Named): IL-12 p70
Target Molecule: Protein
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P43431, P43432

Contents & storage

Antibody pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Store at 2-8°C.

Documents

Manuals & protocols

Recommended products