Oxoid™ CampyGen™ 3.5L Sachet
Thermo Scientific™

Oxoid™ CampyGen™ 3.5L Sachet

Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen 3.5L Sachet.
Have Questions?
Número do catálogoQuantity
CN0035A10/Pk.
Número do catálogo CN0035A
Preço (BRL)
-
Quantity:
10/Pk.

Produce suitable gaseous atmosphere for growth of microaerophilic microorganisms with the Oxoid CampyGen 3.5L Sachet.

  • No water or catalyst required
  • No hydrogen gas produced
  • For use with Thermo Scientific Oxoid 3.5L Anaerobic Jar, Part No. HP0011A, HP0031A
Especificações
Quantity10/Pk.
Volume (Metric)3.5 L
FormatGas Generation Sachet
Product LineThermo Scientific Oxoid
TypeCampyGen Sachet
Unit SizeEach