μPAC™ Trapping Column, 2/pk
μPAC™ Trapping Column, 2/pk
Thermo Scientific™
μPAC™ Trapping Column, 2/pk
Perform large volume (>5 μL) sample injections on capillary and nano-columns with μPAC™ Trapping Columns.
Have Questions?
Número do catálogoPillar Length
COL-TRPNANO16G1B218 μm
Número do catálogo COL-TRPNANO16G1B2
Preço (BRL)
-
Pillar Length:
18 μm
Perform large volume (>5 μL) sample injections on capillary and nano-columns with Thermo Scientific™ μPAC™ Trapping Columns with minimal impact on your total low-flow chromatography analysis time. Their C18 stationary phase support morphology is designed to perfectly match that of capillary and nano- μPAC HPLC columns, with carefully selected dimensions and surface chemistry.
Sample Preconcentration - Preconcentrate samples with very low peptide contents into a sub-microliter trapping volume.
Decreased Sample Loading Times - Depending on the flow rate of the analytical column used, a 10 to 50-fold reduction of the sample loading time can be achieved for high volume (5 to 20 μL) samples.
Sample Clean-up - Effectively remove salts, reagents, and other contaminants that have no affinity for hydrophobic bonded phases, increasing analytical column lifetime and improving spectral quality and detection sensitivity.
Especificações
Bed Length10 mm
Bed Width2 mm
Column TypeMicro Pillar Array
EndcappedYes
Flow Rate10 to 20 μL/min
Interpillar Distance2.5 μm
Max. Pressure350 bar (5076 psi)
Packing MaterialSilicon Chip
Pillar Diameter5 μm
Pillar Length18 μm
Porosity59%
Stationary PhaseC18
Temperature60°C
Pore Size100 to 200 Å
TypeµPAC Trapping Columns
pH1.5 to 7
Unit SizePack of 2