Sterilin™ Cones
Sterilin™ Cones
Thermo Scientific™
Sterilin™ Cones
Safely mount your filter funnels without risk of chipping or slipping. Made of natural rubber, cones are also useful for mounting crucibles onto adapters.
Have Questions?
Catalog NumberDescription
CON0006Set of 6 cones; Assorted
Catalog number CON0006
Price (USD)
-
Request A Quote
Description:
Set of 6 cones; Assorted
Safely mount your filter funnels without risk of chipping or slipping. Thermo Scientific™ Sterilin™ Cones are made of natural rubber, and are also useful for mounting crucibles onto adapters.
  • Set of six cones
  • Manufactured from natural rubber
  • For mounting filter funnels into Büchner flasks
  • Also suitable for mounting Gooch crucibles onto adapters
Ordering Information:
  • Not available in all geographic regions
Specifications
DescriptionSet of 6 cones; Assorted
For Use With (Equipment)Büchner flasks and Gooch crucibles
IncludesSet of six cones
MaterialRubber
Unit SizeEach