Nalgene™ PPCO Centrifuge Bottle,1000mL - FAQs
Nalgene™ PPCO Centrifuge Bottle,1000mL - FAQs
View additional product information for Nalgene™ PPCO Centrifuge Bottle,1000mL - FAQs (CP0610560)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.