Spectra/Mesh Screen Discs
Actual product may vary
Cole-Parmer™
Spectra/Mesh Screen Discs
Four formulations for maximum compatibility Cut the 30 x 30-cm sheets to desired dimensions to fit your needs
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호기공 크기
CP0663530
CP-CP0663530-10/PK으로도 사용됨
100 μm
CP0663100
CP-CP0663100-10/PK으로도 사용됨
5 μm
CP0663106
CP-CP0663106-10/PK으로도 사용됨
53 μm
카탈로그 번호 CP0663530
CP-CP0663530-10/PK으로도 사용됨
제품 가격(KRW)
-
기공 크기:
100 μm
Four formulations for maximum compatibility Cut the 30 x 30-cm sheets to desired dimensions to fit your needs
Four formulations for maximum compatibility Cut the 30 x 30-cm sheets to desired dimensions to fit your needs
사양
직경(미터법)90 mm
유형Spectra/Mesh Screen Disc
물질Nylon
기공 크기100 μm
Unit SizePack of 10