Search
| 카탈로그 번호 | 부피(미터법) | 물질 | 길이(영국식 단위) |
|---|---|---|---|
| CP2555402 CP-CP2555402-1000/CS으로도 사용됨 | 0 to 2000 μL | Borosilicate Glass | 5.71 in. |
| CP2555414 CP-CP2555414-1000/CS으로도 사용됨 | 2.5 mL | Borosilicate Glass | 9 in. |
These Pasteur pipettes are manufactured to conform to rigid specifications and are suitable for use in hematology, blood banks, bacteriology, and general laboratory applications. These economical pipettes feature straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment. Pipettes are packed in cartons of 250, four cartons per case of 1000. Please order bulbs separately.