ChargeSwitch® Purification Buffer (N5) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: CS11210

Invitrogen™  Related applications: DNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ChargeSwitch® Forensic DNA Purification Kit is a magnetic bead method designed to accommodate all forensic crime scene sample types for STR profiling. The kit is scalable for single-tube use or for automated extraction.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer Type: Binding Buffer
Product Line: ChargeSwitch®
Product Size: 300 mL
Green Features: Fewer resources used, Less hazardous, Less waste
Shipping Condition: Room Temperature

Contents & storage

All components of the ChargeSwitch® Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly."

Documents

Manuals & protocols