ChargeSwitch™ gDNA Rendered Meat Purification Kit
Invitrogen™

ChargeSwitch™ gDNA Rendered Meat Purification Kit

The ChargeSwitch™ gDNA Rendered Meat Purification Kit allows for rapid and efficient purification of genomic DNA (gDNA) from animal feed, animal meal, and heparin products for downstream molecular analysis.
Have Questions?
Catalog NumberQuantity
CS40010050 reactions
Catalog number CS400100
Price (USD)
-
Quantity:
50 reactions

The ChargeSwitch™ gDNA Rendered Meat Purification Kit allows for rapid and efficient purification of genomic DNA (gDNA) from animal feed, animal meal, and heparin products for downstream molecular analysis. The kit is listed in the U.S. Food and Drug Administration Center for Veterinary Medicine's protocols as a sample preparation method suitable for monitoring crude heparin quality and testing animal feed for banned materials such as specific animal proteins.

Note: The number of reactions possible with this kit is dependent upon the starting sample type: animal feed and meal samples = 50 preparations, heparin products = 25 preparations.

The ChargeSwitch™ gDNA Rendered Meat Purification Kit protocol starts with prepared lysates and takes approximately 15 minutes to complete without centrifugation, vacuum, or organic solvents (for batches of up to 5 samples per operator—larger batch sizes will require longer processing times). The kit employs a magnetic bead with a switchable surface. Low pH conditions cause binding of DNA to the bead while high pH conditions cause the release of the DNA.

Tested by the U.S. Food and Drug Administration

The ChargeSwitch™ gDNA Rendered Meat Purification Kit has been thoroughly tested by the U.S. Food and Drug Administration for processing up to 500 mg of animal feed, 250 mg of animal meal, and 250 mg of heparin product per reaction.

References

Development of a Multiplex Real-Time PCR Assay for the Detection of Ruminant DNA in Raw Materials used for Monitoring Crude Heparin for Quality: www.fda.gov/AnimalVeterinary/ScienceResearch/ToolsResources/ucm350289.html

Development, evaluation, and peer verification of a rapid real-time PCR method for the detection of animal material. Journal of Food Protection, Vol. 72, No. 11, 2009, Pages 2368–2374. U.S. Food and Drug Administration, Center for Veterinary Medicine, Office of Research, Laurel, Maryland 20708, USA.

Specifications
For Use With (Application)Real-Time Quantitative PCR (qPCR), PCR
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions50 Reactions
Product LineChargeSwitch
Quantity50 reactions
Sample TypeBiopharmaceutical Production Samples, Food and Environmental
Shipping ConditionRoom Temperature
TargetGenomic DNA
Test Time15 min.
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
• Magnetic Beads, 1.1 mL
• SDS Solution, 10 mL
• 10% Detergent, 10 mL
• Lysis Buffer, 100 mL
• Precipitation Buffer, 38.5 mL
• Wash Buffer, 200 mL
• Elution Buffer, 15 mL