IL-2 Porcine Matched Antibody Pair

Catalog number:  CSC1243

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This Antibody Pair Kit comes with matched antibody pairs to detect and quantify protein levels of swine IL-2. Also available is Buffer Kit for Antibody Pairs that saves time with premade, easy-to-use buffers and solutions optimized for use with Invitrogen Antibody Pairs. The Assay Buffer can be used as a blocking reagent for ELISA plates as well as a diluent for ELISA standards and samples, detection antibody, and HRP conjugate. Buffer reagents needed to complete the reaction are sold separately under Catalog No. CNB0011.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Range: See product documentation
Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Novex
Gene Aliases: IL-2;,TCGF, POIL2
Gene ID: 396868
Gene Name: interleukin 2
Gene Symbol: IL2
Immunoassay Kit Format: Matched Antibody Pair
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 10 x 96 tests
Protein Aliases: IL-2, T-cell growth factor, TCGF
Protein Name: Interleukin-2
Shipping Condition: Wet Ice
Target (Kit Named): IL-2
Target Molecule: Protein
Target Species (Named): Porcine
Target Species (Validated): Porcine

Contents & storage

Antibody pair Kit contains matched, pre-titred and fully optimized Capture (coating) and Detection Antibodies, 3 vials of Recombinant Standard, Streptavidin-HRP concentrate and technical data sheet.
Store at 2-8°C.

Documents

Manuals & protocols

Recommended products