Interested in a bulk or custom version of this product? Request pricing.

XRCC5 AND XRCC6 Recombinant Human Protein, N-His Tag

Catalog number: CT018H07B50

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

X-ray Repair Complementing defective repair in Chinese hamster cells 5 and 6 Heterodimer (XRCC5 and XRCC6), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the XRCC5 (P13010) (Met 1-Ile 732) fused to a polyhistidine tag at the N-terminus, constructed the plasmid 1, and from a DNA sequence encoding the XRCC6 (P12956) (Met 1-Asp 609) fused to a polyhistidine tag at the N-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.

Activity: This recombinant protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 8.0.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biologica Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Other Research
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: KARP-1, KARP1, KU80, KUB2, Ku86, NFIV, CTA-216E10.7, CTC75, CTCBF, G22P1, KU70, ML8, TLAA
Gene ID: 7520 and 2547
Gene Symbol: XRCC5 AND XRCC6
Molecular Weight: 157 kDa
Product Line: Sino Biological®
Product Size: 50 µg
Protein Form: Full Length, Recombinant
Protein Name: X-Ray Repair Complementing Defective Repair In Chinese Hamster Cells 5 And 6 Heterodimer
Protein Tag or Fusion: N-His
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 1 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols