Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 μg
Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 μg
Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 μg
Thermo Scientific™
Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks, 200 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Catalog NumberConcentration
CT0758B200 μg
Catalog number CT0758B
Price (GBP)
-
Concentration:
200 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Fosfomycin Antimicrobial Susceptibility Disks.
The disc contains 200μg of the active ingredient, fosfomycin, with the addition of 50μg of glucose-6-phospate, as required by the EUCAST methodology. Discs must be used for E.coli only and for other Enterobacterales and Staphylococcus spp., an MIC method must be used.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Antibiotic CodeFOT
Antimicrobial AgentsFosfomycin
Concentration200 μg
DescriptionOxoid Fosfomycin Antimicrobial Susceptibility Disks
No. per Pack5 Cartridges/Pk.
Packaging Type50 Disks/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach