Oxoid™ Telithromycin Antimicrobial Susceptibility Disks, 15 μg
Oxoid™ Telithromycin Antimicrobial Susceptibility Disks, 15 μg
Oxoid™ Telithromycin Antimicrobial Susceptibility Disks, 15 μg
Thermo Scientific™
Oxoid™ Telithromycin Antimicrobial Susceptibility Disks, 15 μg
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Have Questions?
Número do catálogoConcentration
CT1714B15 μg
Número do catálogo CT1714B
Preço (BRL)
-
Concentration:
15 μg
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Telithromycin Antimicrobial Susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Especificações
Antibiotic CodeTEL
Antimicrobial AgentsTelithromycin
Certifications/ComplianceComplies with CLSI, BSAC, SFM guidelines
Concentration15 μg
DescriptionOxoid Telithromycin Antimicrobial Susceptibility Disks
No. per Pack5 Cartridges/Pk.
Packaging Type50 Disks/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit SizeEach