Oxoid™ Cefonicid Antimicrobial Susceptibility discs
Oxoid™ Cefonicid Antimicrobial Susceptibility discs
Oxoid™ Cefonicid Antimicrobial Susceptibility discs
Oxoid™ Cefonicid Antimicrobial Susceptibility discs
Thermo Scientific™
Oxoid™ Cefonicid Antimicrobial Susceptibility discs
This product has been discontinued. Please refer to our AST disc range for a suitable alternative.
Have Questions?
Catalog NumberConcentration
CT1894B30 μg
Catalog number CT1894B
Price (USD)
-
Request A Quote
Concentration:
30 μg
Thermo Scientific™ Oxoid™ Cefonicid Antimicrobial Susceptibility disc CT1894B Cefonicid 30μg (CID30) has been withdrawn from the range as of April 2015. Please refer to our AST disc range for a suitable alternative.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Antibiotic CodeCID
Antimicrobial AgentsCefonicid
Certifications/ComplianceComplies with CLSI guidelines
Concentration30 μg
DescriptionOxoid Cefonicid Antimicrobial Susceptibility Discs
No. per Pack5 Cartridges/Pk.
Packaging Type50 Discs/Cartridge
Quantity50 Discs/Cartridge
Product TypeAntimicrobial Susceptibility Discs
Test TypeAntimicrobial Susceptibility
Unit Size-