BioHale™ Sucrose
DFE Pharma

BioHale™ Sucrose

BioHale Sucrose is a non-reducing sugar that does not react with amino acids or proteins, inhibiting the Maillard reaction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
D11822241 kg
D117877120 kg
Catalog number D1182224
Price (MXN)
-
Quantity:
1 kg

BioHale Sucrose is a non-reducing sugar that does not react with amino acids or proteins, inhibiting the Maillard reaction. BioHale Sucrose provides solution-state stabilization to fragile biomolecules.

Cryo- and lyo-protectant

BioHale Sucrose, a high-purity disaccharide excipient, protects biologic drugs from the freeze related (cryoprotectant) and drying related (lyoprotectant) stresses. This makes BioHale Sucrose particularly suitable in the stabilization process of today's biologics.

Quality

  • High-purity, low-endotoxin grade produced by active purification process
  • Manufactured in The Netherlands, Europe
  • FDA inspected and state-of-the-art cGMP facility
  • Multi-compendial specification complies with Ph. Eur., USP-NF, JP, ChP
  • Chinese DMF available
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS57-50-1
Chemical Name or MaterialSucrose
ColorWhite to Off-White
DOT InformationNot subject to limitations due to transport regulations.
EINECS Number200-334-9
For Use With (Application)Stabilizing biologics for use in parenteral, ophthalmic, or inhalation application, Lyophilization
Melting Point186°C
Molecular FormulaC12H22O11
PackagingHDPE container with PE inner liner
RTECS NumberWN6500000
Shelf Life36 Months
Storage Note 1No special/controlled storage conditions required
SynonymSaccharose, Sugar beet
GradeMulti-Compendial
Molecular Weight (g/mol)342.3
Percent Purity98-102%
Physical FormCrystalline Powder
Quantity1 kg
pH5.5 to 7.5 (342 g/L, H2O, 25°C)
Unit SizeEach