Tyvek™ 500C Apron
DuPont™
Tyvek™ 500C Apron
Shin-length apron with neck and waist ties for enhanced protection of body parts that are more exposed to hazardous substances. Constructed of flash spun, high density polyethylene for an ideal balance of protection, durability, and comfort.
Have Questions?
Número de catálogo D13396088
Precio (USD)
-
Shin-length apron with neck and waist ties for enhanced protection of body parts that are more exposed to hazardous substances. Constructed of flash spun, high density polyethylene for an ideal balance of protection, durability, and comfort.
  • Enhanced protection for body parts that are more exposed to hazardous substances
  • Composed of flash spun, high density polyethylene for an ideal balance of protection, durability, and comfort
  • Ties at the waist and ankles
  • Shin-length
Especificaciones
Descripción del artículoShin Length Apron with Ties
Longitud (imperial)42.5 in.
Longitud (métrico)108 cm
Tipo de costuraSeamless
Tipo de tapónTies
ColorWhite
DesechablesSingle-use
MaterialPolyethylene
TamañoUniversal
EsterilidadNon-sterile
Unit SizeEach