DRAQ7 DROP & GO™ Dye
Invitrogen™

DRAQ7 DROP & GO™ Dye

DRAQ7 DROP & GO Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry.Read more
Have Questions?
Catalog NumberQuantity
D1510790 assays
Catalog number D15107
Price (USD)
372.65
Online exclusive
497.00
Save 124.35
90 assays
Quantity:
90 assays
Price (USD)
372.65
Online exclusive
497.00
Save 124.35
90 assays
DRAQ7 DROP & GO Dye is a bright, easy-to-use fluorescent DNA dye used to exclude non-viable cells by flow cytometry. This dye has excitation maxima at 599/644 nm and emission maxima at 678/697 nm.

• Stable at room temperature
• Convenient, ready-to-use dropper-bottle format—no need to dilute, weigh, or pipette
• No UV excitation and no emission spectral overlap with PE

DRAQ7 Dye is a membrane-impermeable dye that rapidly stains the double-stranded DNA (dsDNA) of dead or permeabilized cells. The dye can be excited by wavelengths from 488 to 647 nm, though it is optimally excited by red laser lines. Its emission maxima is at 697 nm when intercalated with dsDNA.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity90 assays
Product TypeDye
Unit Size90 assays
Contents & Storage
3 x 2.5 mL

Citations & References (1)

Citations & References
Abstract
Isolation of photoreceptors from mature, developing, and regenerated zebrafish retinas, and of microglia/macrophages from regenerating zebrafish retinas.
Authors:Sun C, Mitchell DM, Stenkamp DL,
Journal:Exp Eye Res
PubMed ID:30096325
This paper describes experimental procedures for the dissociation of retinal cells of the zebrafish (Danio rerio) for subsequent fluorescence-activated cell sorting (FACS) and gene expression studies. Methods for dissociation of zebrafish retinas followed by FACS and RNA isolation were optimized. This methodology was applied to isolate pure sorted samples of ... More