Dextran, Amino, 10,000 MW
Invitrogen™

Dextran, Amino, 10,000 MW

Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of liveRead more
Have Questions?
Catalog NumberQuantity
D18601 g
Catalog number D1860
Price (USD)
249.65
Online Exclusive
268.00
Save 18.35 (7%)
Each
Quantity:
1 g
Price (USD)
249.65
Online Exclusive
268.00
Save 18.35 (7%)
Each
Labeled dextrans are hydrophilic polysaccharides most commonly used in microscopy studies to monitor cell division, track the movement of live cells, and to report the hydrodynamic properties of the cytoplasmic matrix. The labeled dextran is commonly introduced into the cells via microinjection.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

Dextran Specifications:

Label (Ex/Em): None (can be conjugated to dyes and other molecules using amine-reactive moities)
Size: 10,000 MW

High Manufacturing Standards of Molecular Probes™ Dextrans
We offer more than 50 fluorescent and biotinylated dextran conjugates in several molecular weight ranges. Dextrans are hydrophilic polysaccharides characterized by their moderate-to-high molecular weight, good water solubility, and low toxicity. They also generally exhibit low immunogeniticy. Dextrans are biologically inert due to their uncommon poly-(α-D-1,6-glucose) linkages, which render them resistant to cleavage by most endogenous cellular glycosidases.

In most cases, Molecular Probes™ fluorescent dextrans are much brighter and have higher negative charge than dextrans available from other sources. Furthermore, we use rigorous methods for removing as much unconjugated dye as practical, and then assay our dextran conjugates by thin-layer chromatography to help ensure the absence of low molecular weight contaminants.

A Wide Selection of Substituents and Molecular Weights
Molecular Probes™ dextrans are conjugated to biotin or a wide variety of fluorophores, including seven of our Alexa Fluor™ dyes (Molecular Probes dextran conjugates–Table 14.4) and are available in these nominal molecular weights (MW): 3,000; 10,000; 40,000; 70,000; 500,000; and 2,000,000 daltons.

Dextran Net Charge and Fixability
We employ succinimidyl coupling of our dyes to the dextran molecule, which, in most cases, results in a neutral or anionic dextran. The reaction used to produce the Rhodamine Green™ and Alexa Fluor 488 dextrans results in the final product being neutral, anionic, or cationic. The Alexa Fluor, Cascade Blue, lucifer yellow, fluorescein, and Oregon Green dextrans are intrinsically anionic, whereas most of the dextrans labeled with the zwitterionic rhodamine B, tetramethylrhodamine, and Texas Red™ dyes are essentially neutral. To produce more highly anionic dextrans, we have developed a proprietary procedure for adding negatively charged groups to the dextran carriers; these products are designated “polyanionic” dextrans.

Some applications require that the dextran tracer be treated with formaldehyde or glutaraldehyde for subsequent analysis. For these applications, we offer “lysine-fixable” versions of most of our dextran conjugates of fluorophores or biotin. These dextrans have covalently bound lysine residues that permit dextran tracers to be conjugated to surrounding biomolecules by aldehyde-mediated fixation for subsequent detection by immunohistochemical and ultrastructural techniques. We have also shown that all of our 10,000 MW Alexa Fluor dextran conjugates can be fixed with aldehyde-based fixatives.

Key Applications Using Labeled Dextrans
There are a multitude of citations describing the use of labeled dextrans. Some of the most common uses include:

Neuronal tracing (anterograde and retrograde) in live cells
Cell lineage tracing in live cells
Neuroanatomical tracing
Examining intercellular communications (e.g., in gap junctions, during wound healing, and during embryonic development)
Investigating vascular permeability and blood–brain barrier integrity
Tracking endocytosis
Monitoring acidification (some dextran–dye conjugates are pH-sensitive)
Studying the hydrodynamic properties of the cytoplasmic matrix

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeUnlabeled
Product TypeDextran
Quantity1 g
Shipping ConditionRoom Temperature
Product LineInvitrogen
Unit SizeEach
Contents & Storage
Store at room temperature.

Citations & References (7)

Citations & References
Abstract
Monitoring the permeability of the nuclear envelope during the cell cycle.
Authors:Lénárt P, Ellenberg J,
Journal:Methods
PubMed ID:16343937
'In animal organisms the nuclear envelope (NE) dis-assembles during cell division resulting in complete intermixing of cytoplasmic and nuclear compartments. This leads to the activation of many mitotic enzymes, which were kept away from their substrates or regulators by nuclear or cytoplasmic sequestration in interphase. Nuclear envelope breakdown (NEBD) is ... More
Fluorescence correlation microscopy of cells in the presence of autofluorescence.
Authors:Brock R, Hink MA, Jovin TM
Journal:Biophys J
PubMed ID:9788950
'Fluorescence correlation microscopy (FCM), the combination of fluorescence correlation spectroscopy (FCS) and digital microscopy (Brock and Jovin, 1998. Cell. Mol. Biol. 44:847-856), has been implemented for measuring molecular diffusion and association in living cells with explicit consideration of autocorrelations arising from autofluorescence. Autofluorescence excited at 532 nm colocalizes with mitochondria, ... More
Localized calcium signals along the cleavage furrow of the Xenopus egg are not involved in cytokinesis.
Authors:Noguchi T, Mabuchi I
Journal:Mol Biol Cell
PubMed ID:11950937
It has been proposed that a localized calcium (Ca) signal at the growing end of the cleavage furrow triggers cleavage furrow formation in large eggs. We have examined the possible role of a Ca signal in cleavage furrow formation in the Xenopus laevis egg during the first cleavage. We were ... More
Mycobacterium avium- and Mycobacterium tuberculosis-containing vacuoles are dynamic, fusion-competent vesicles that are accessible to glycosphingolipids from the host cell plasmalemma.
Authors:Russell DG, Dant J, Sturgill-Koszycki S
Journal:J Immunol
PubMed ID:8648123
The vacuoles inhabited by viable Mycobacterium avium and Mycobacterium tuberculosis show limited fusion with endosomal and lysosomal compartments. This ability to regulate the maturation of their phagosomal compartments and restrict their differentiation into hydrolytically active vacuoles appears to correlate with the survival of the bacilli. Data presented in this current ... More
Imaging of endosome fusion in BHK fibroblasts based on a novel fluorimetric avidin-biotin binding assay.
Authors:Emans N, Biwersi J, Verkman AS
Journal:Biophys J
PubMed ID:8527685
A fluorescence assay of in vivo endosome fusion was developed and applied to define the kinetics of endosome fusion in baby hamster kidney (BHK) fibroblasts. The assay is based on an approximately 10-fold enhancement of the green fluorescence of BODIPY-avidin upon biotin binding. The BODIPY-avidin fluorescence enhancement occurred in < ... More