Owl™ D2 Wide-Gel Electrophoresis System
Thermo Scientific™

Owl™ D2 Wide-Gel Electrophoresis System

Run 10 to 80 samples on one gel with this wide-gel electrophoresis system.
Have Questions?
Catalog NumberType
D2-BPWide w/Buffer Exchange Ports
Catalog number D2-BP
Price (USD)
-
Type:
Wide w/Buffer Exchange Ports
Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.
  • Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
  • System is excellent for screening PCR samples or routine DNA fragment analysis.
  • Small footprint saves space on lab bench.
  • Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Electrode placement allows gel to be run quickly without compromising resolution.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • EasyCast gasketed UVT gel tray
  • External multiple gel caster
  • Four combs (two 30-well and two 40-well, 1.5mm thick)
Specifications
Certifications/ComplianceCE marked
DescriptionComplete System with Buffer Exchange Ports
Dimensions (L x W x H)7.38 x 7.75 x 4 in. (19 x 20 x 10 cm)
Height (English)4 in.
Height (Metric)10 cm
Length (English)7.38 in.
Length (Metric)19 cm
Length (Metric) Gel14.4 cm
Sample Size10 to 80
Volume (Metric) Buffer600 mL
Width (English)7.75 in.
Width (English) Gel6.5
Width (Metric)20 cm
Width (Metric) Gel10.2 cm
Max. Voltage150 V
Product LineOwl
TypeWide w/Buffer Exchange Ports
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.