Owl™ D2 Wide-Gel System Replacement Parts
Actual product may vary
Thermo Scientific™
Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D2-CSTCaster
D2-GKGasket
Catalog number D2-CST
Price (USD)
173.70
Online Exclusive
193.00
Save 19.30 (10%)
Each
Type:
Caster
Price (USD)
173.70
Online Exclusive
193.00
Save 19.30 (10%)
Each
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
DescriptionExternal caster (no trays/combs)
For Use With (Equipment)Owl™ D2 Wide Gel Electrophoresis System
Product LineOwl
TypeCaster
Unit SizeEach