BODIPY™ FL NHS Ester (Succinimidyl Ester)
Invitrogen™

BODIPY™ FL NHS Ester (Succinimidyl Ester)

BODIPY™ FL dye is bright, green fluorescent dye with similar excitation and emission to fluorescein (FITC) or Alexa Fluor™ 488Read more
Have Questions?
Catalog NumberQuantity
D21845 mg
Catalog number D2184
Price (USD)
592.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
592.00
Each
BODIPY™ FL dye is bright, green fluorescent dye with similar excitation and emission to fluorescein (FITC) or Alexa Fluor™ 488 dye. It has a high extinction coefficient and fluorescence quantum yield and is relatively insensitive to solvent polarity and pH change. In contrast to the highly water soluble fluorophores Alexa Fluor™ 488 dye and fluorescein (FITC), BODIPY™ dyes have unique hydrophobic properties ideal for staining lipids, membranes, and other lipophilic compounds. BODIPY™ FL dye has a relatively long excited-state lifetime (typically 5 nanoseconds or longer), which is useful for fluorescence polarization-based assays and a large two-photon cross-section for multiphoton excitation. In addition to reactive dye formulations, we offer BODIPY™ FL dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of BODIPY™ FL is the most popular tool for conjugating the dye to a protein or antibody. NHS esters can be used to label the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting BODIPY™ FL conjugates exhibit bright fluorescence, narrow emission bandwidths, and relatively long excited-state lifetimes, which can be useful for fluorescence polarization assays and two-photon excitation (TPE) microscopy.

This reactive dye contains a C3 alkyl spacer between the fluorophore and the NHS ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.

Detailed information about this BODIPY™ FL NHS ester:

Fluorophore label: BODIPY™ FL dye
Reactive group: NHS ester (succinimidyl ester)
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 502/510 nm
Extinction coefficient: 82,000 cm-1M-1
Molecular weight: 389.16

Typical Conjugation Reaction
Amine-reactive reagents can be conjugated with virtually any protein or peptide; the provided protocol is optimized for IgG antibodies. The reaction can be scaled for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The BODIPY™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO), and the reaction is carried out in 0.1-0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free BODIPY™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration medium with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission510 nm
Excitation502 nm
Label or DyeBODIPY™ FL
Product TypeNHS Ester (Succinimidyl Ester)
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeBODIPY Dyes
Product LineBODIPY
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (93)

Citations & References
Abstract
Authors:
Journal:
PubMed ID:16184172
Synthesis and properties of fluorescent beta-adrenoceptor ligands.
Authors:Heithier H, Hallmann D, Boege F, Reiländer H, Dees C, Jaeggi KA, Arndt-Jovin D, Jovin TM, Helmreich EJ
Journal:Biochemistry
PubMed ID:7914090
We describe the synthesis of bordifluoropyrromethene (BODIPY), fluorescein, and related fluorescent derivatives of the beta-adrenergic ligand CGP 12177. With these probes we screened insect (Sf9) cells stably transformed with the human beta 2-adrenoceptor gene and expressing (2-3.5) x 10(5) human beta 2-adrenoceptors per cell. Among these derivatives only BODIPY-CGP gave ... More
LDL-mediated interaction of Lp[a] with HepG2 cells: a novel fluorescence microscopy approach.
Authors:Hofer G, Steyrer E, Kostner GM, Hermetter A
Journal:J Lipid Res
PubMed ID:9458265
'We studied the topography of Lp[a]-LDL-cell interactions by means of fluorescence microscopy, using fluorescence-labeled lipoproteins. In contrast to known methods which are based on noncovalent labeling of lipoproteins by positively charged amphiphiles, the protein moiety of LDL and Lp[a] was covalently labeled with either BODIP-succinimide-ester (green) or rhodamine X iodoacetamide ... More
Interaction of a small heat shock protein of the fission yeast, Schizosaccharomyces pombe, with a denatured protein at elevated temperature.
Authors:Hirose M, Tohda H, Giga-Hama Y, Tsushima R, Zako T, Iizuka R, Pack C, Kinjo M, Ishii N, Yohda M
Journal:J Biol Chem
PubMed ID:16055437
'We have expressed, purified, and characterized one small heat shock protein of the fission yeast Schizosaccharomyces pombe, SpHsp16.0. SpHsp16.0 was able to protect citrate synthase from thermal aggregation at 45 degrees C with high efficiency. It existed as a hexadecameric globular oligomer near the physiological growth temperature. At elevated temperatures, ... More
A comparison of the emission efficiency of four common green fluorescence dyes after internalization into cancer cells.
Authors:Hama Y, Urano Y, Koyama Y, Bernardo M, Choyke PL, Kobayashi H
Journal:Bioconjug Chem
PubMed ID:17105220
'In vivo optical imaging to enhance the detection of cancer during endoscopy or surgery requires a targeted fluorescent probe with high emission efficiency and high signal-to-background ratio. One strategy to accurately detect cancers is to have the fluorophore internalize within the cancer cells permitting nonbound fluorophores to be washed away ... More