Die ratiometrische mitochondriale Membranpotentialsonde JC-9 weist eine grüne Fluoreszenz auf, die unabhängig vom Membranpotential ist und eine Zunahme der rotenWeitere Informationen
Have Questions?
Katalognummer
Menge
D22421
5 mg
Katalognummer D22421
Preis (EUR)
924,00
Each
Menge:
5 mg
Preis (EUR)
924,00
Each
Die ratiometrische mitochondriale Membranpotentialsonde JC-9 weist eine grüne Fluoreszenz auf, die unabhängig vom Membranpotential ist und eine Zunahme der roten Fluoreszenz bei hyperpolarisierten Membranpotentialen aufweist.
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
NachweisverfahrenFluoreszent
Menge5 mg
VersandbedingungRaumtemperatur
Subzelluläre LokalisationMitochondrien
FarbeGrün
Zur Verwendung mit (Geräte)Fluoreszenzmikroskop, Durchflusszytometer
ProdukttypMitochondrienmembranpotential-Sonde
Unit SizeEach
Inhalt und Lagerung
Lagerung bei Raumtemperatur und vor Licht geschützt
Häufig gestellte Fragen (FAQ)
I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?
If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).
What is the difference between fast and slow-response membrane potential probes?
Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.
What type of membrane potential indicators do you offer and how should I choose one for my experiment?
A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).
'Late-stage malignant melanoma is a cancer that is refractory to current chemotherapeutic treatments. The average survival time for patients with such a diagnosis is 6 months. In general, the vast majority of anticancer drugs operate through induction of cell cycle arrest and cell death in either the DNA synthesis (S) ... More
Chlorophenols and chlorocatechols induce apoptosis in human lymphocytes (in vitro).
Authors:Michalowicz J, Sicinska P,
Journal:Toxicol Lett
PubMed ID:19766705
In this work the effect of 2,4,5-trichlorophenol (2,4,5-TCP), pentachlorophenol (PCP), 4,6-dichloroguaiacol (4,6-DCG), tetrachloroguaiacol (TeCG), 4,5-dichlorocatechol (4,5-DCC) and tetrachlorocatechol (TeCC) on the induction of apoptosis in human peripheral blood lymphocytes was examined. The analysis of the changes in mitochondrial transmembrane potential (DeltaPsi(m)) was performed using JC-9 fluorescent probe. It was noted ... More
Markers of apoptosis: methods for elucidating the mechanism of apoptotic cell death from the nervous system.
Authors:Smyth PG, Berman SA,
Journal:Biotechniques
PubMed ID:11911667
Apoptosis is a highly conserved energy-requiring program for non-inflammatory cell death that is important in both normal physiology and disease. Numerous techniques have been used to study apoptosis. In the nervous system, apoptosis is necessary for normal development, but it also occurs in many acute and chronic pathologic conditions. This ... More
DGAT1-Dependent Lipid Droplet Biogenesis Protects Mitochondrial Function during Starvation-Induced Autophagy.
Authors:
Journal:Dev Cell
PubMed ID:28697336
A role for mitochondria in the establishment and maintenance of the maize root quiescent center.