DiOC6(3) (3,3'-Dihexyloxacarbocyanine Iodide)
Invitrogen™
DiOC6(3) (3,3'-Dihexyloxacarbocyanine Iodide)
DiOC6(3) es un colorante lipofílico, con permeablidad celular, verde fluorescente que es selectivo para la mitocondria de las células vivasMás información
Have Questions?
Número de catálogoCantidad
D273100 mg
Número de catálogo D273
Precio (MXN)
-
Cantidad:
100 mg
DiOC6(3) es un colorante lipofílico, con permeablidad celular, verde fluorescente que es selectivo para la mitocondria de las células vivas cuando se utiliza a bajas concentraciones. A concentraciones más altas, el tinte se puede usar para teñir otras membranas internas, como el retículo endoplásmico.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Método de detecciónFluorescente
Cantidad100 mg
Condiciones de envíoTemperatura ambiente
Localización subcelularMembranas celulares y lípidos
ColorVerde
Para utilizar con (equipo)Microscopio de fluorescencia
Tipo de productoDiOC6(3)
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente y proteger de la luz.
Preguntas frecuentes
I stained my cells with a lipophilic cyanine dye, like DiI, but the signal was lost when I tried to follow up with antibody labeling. Why?
Since these dyes insert into lipid membranes, any disruption of the membranes leads to loss of the dye. This includes permeabilization with detergents like Triton X-100 or organic solvents like methanol. Permeabilization is necessary for intracellular antibody labeling, leading to loss of the dye. Instead, a reactive dye such as CFDA SE should be used to allow for covalent attachment to cellular components, thus providing for better retention upon fixation and permeabilization.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How long does it take for lipophlic tracers to transport along the membrane? How much faster are the FAST lipophilic dyes?
The transport is fairly slow, around 6 mm/day in live tissue and slower in fixed tissue, so diffusion of lipophilic carbocyanine tracers from the point of their application to the terminus of a neuron can take several days to weeks The FAST DiO and DiI analogs (which have unsaturated alkyl tails) can improve transport rate by around 50%.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Which form of the lipophilic tracers (DiO, DiI, DiD, etc) should I use?
Select the dye that is compatible with your available excitation source(s) and emission filter set/channels. The solid, paste and crystal forms can be applied directly to neurons in tissues. For labeling cells in culture or microinjection, the lipophilic dyes in solution or solid form can be used.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I want to label two cell populations and then perform a cell fusion assay. Which reagents are best for imaging this?
Lipophilic cyanine dyes are preferred for this sort of assay, since they insert into cellular membranes and then, upon fusion, are shared by the fused cells as the membranes are shared. For example, one cell population can be labeled with DiI (orange-red) and another cell population can be labeled with DiO (green), and when the cells fuse, the combined color appears yellow (when imaged with a dual-bandpass filter set).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (388)
Citations & References
Abstract
Authors:
Journal:
PubMed ID:10891486
The mitochondrial death/life regulator in apoptosis and necrosis.
Authors:Kroemer G,Dallaporta B,Resche-Rigon M
Journal:Annual review of physiology
PubMed ID:9558479
Both physiological cell death (apoptosis) and, in some cases, accidental cell death (necrosis) involve a two-step process. At a first level, numerous physiological and some pathological stimuli trigger an increase in mitochondrial membrane permeability. The mitochondria release apoptogenic factors through the outer membrane and dissipate the electrochemical gradient of the ... More
Alcohol-induced thymocyte apoptosis is accompanied by impaired mitochondrial function.
Authors:Wang JF,Spitzer JJ
Journal:Alcohol (Fayetteville, N.Y.)
PubMed ID:9014030
This study examines the effects of chronic alcohol consumption on thymic apoptosis with or without pretreatment with E. coli lipopolysaccharide (LPS). Apoptotic cell death of thymocytes was monitored by DNA fragments in gel electrophoresis and the appearance of apoptotic cells by flow cytometry. Changes in mitochondrial membrane potential (MMP), as ... More
The dynamin-related GTPase, Dnm1p, controls mitochondrial morphology in yeast.
Authors:Otsuga D, Keegan BR, Brisch E, Thatcher JW, Hermann GJ, Bleazard W, Shaw JM
Journal:J Cell Biol
PubMed ID:9786946
'The Saccharomyces cerevisiae Dnm1 protein is structurally related to dynamin, a GTPase required for membrane scission during endocytosis. Here we show that Dnm1p is essential for the maintenance of mitochondrial morphology. Disruption of the DNM1 gene causes the wild-type network of tubular mitochondrial membranes to collapse to one side of ... More
Involvement of endonuclease G in nucleosomal DNA fragmentation under sustained endogenous oxidative stress.
Authors:Ishihara Y, Shimamoto N
Journal:J Biol Chem
PubMed ID:16407272
'We have previously shown that inhibition of catalase and glutathione peroxidase activities by 3-amino-1,2,4-triazole (ATZ) and mercaptosuccinic acid (MS), respectively, in rat primary hepatocytes caused sustained endogenous oxidative stress and apoptotic cell death without caspase-3 activation. In this study, we investigated the mechanism of this apoptotic cell death in terms ... More