Cellulose Dialysis Tubing Strips
Actual product may vary

Cellulose Dialysis Tubing Strips

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
Have Questions?
Catalog NumberMWCOCapacity (Metric)
D3-143.5 kDa3.57 mL/cm
Catalog number D3-14
HSN 90272000
Price (INR)
169,148.00
Each
MWCO:
3.5 kDa
Capacity (Metric):
3.57 mL/cm
Price (INR)
169,148.00
Each
  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 305mm strips
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14, 000Da, 8, 000Da, and 3, 500Da molecular weight cut offs
Specifications
Product TypeDialysis Tubing Strips
MWCO3.5 kDa
MaterialCellulose
Diameter (Metric)21.3 mm
Length (Metric)30.5 m
Capacity (Metric)3.57 mL/cm
ColorTransparent
Quantity1 Each
For Use With (Application)For life sciences research
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.