Owl™ D3-14 System Replacement Parts
Actual product may vary
Thermo Scientific™
Owl™ D3-14 System Replacement Parts
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts including UVT gel tray, gaskets and external caster.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D3-UVT-14Gel Tray
D3-CST-14Caster
D3-GKGasket
Catalog number D3-UVT-14
Price (USD)
-
Type:
Gel Tray
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to enable up to 12 sample sets on the same gel
Specifications
DescriptionEasyCast™ gasketed UVT Gel Tray
Length (Metric)14 cm
Width (Metric)23 cm
For Use With (Equipment)Owl™ D3 Series Horizontal Electrophoresis System
Product LineOwl
TypeGel Tray
Unit SizeEach