BODIPY&trade; FL DHPE (<i>N</i>-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene-3-Propionyl)-1,2-Dihexadecanoyl-<i>sn</i>-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
Invitrogen™

BODIPY™ FL DHPE (N-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionyl)-1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)

BODIPY™ DHPEは、リン脂質の頭部基に緑色蛍光色素が結合しています。BODIPY™ FL™は高濃度でエキシマーを形成できます。また、ピレン、DPH、NBDなどのフルオロフォアとは異なり、環境に対して比較的低感度で詳細を見る
製品番号(カタログ番号)数量
D3800100 μg
製品番号(カタログ番号) D3800
価格(JPY)
35,100
キャンペーン価格
Ends: 27-Mar-2026
58,500
割引額 23,400 (40%)
Each
数量:
100 μg
BODIPY™ DHPEは、リン脂質の頭部基に緑色蛍光色素が結合しています。BODIPY™ FL™は高濃度でエキシマーを形成できます。また、ピレン、DPH、NBDなどのフルオロフォアとは異なり、環境に対して比較的低感度で、水性環境と脂質環境の両方で蛍光を発します。
研究用にのみ使用できます。診断用には使用いただけません。
仕様
化学物質名または材質Phospholipids
推奨保存方法Store in freezer (-5 to -30°C) and protect from light.
物理的フォームSolid
製品ラインBODIPY
数量100 μg
Unit SizeEach

引用および参考文献 (10)

引用および参考文献
Abstract
Cell-free reconstitution of vacuole membrane fragmentation reveals regulation of vacuole size and number by TORC1.
Authors:Michaillat L, Baars TL, Mayer A,
Journal:Mol Biol Cell
PubMed ID:22238359
'Size and copy number of organelles are influenced by an equilibrium of membrane fusion and fission. We studied this equilibrium on vacuoles-the lysosomes of yeast. Vacuole fusion can readily be reconstituted and quantified in vitro, but it had not been possible to study fission of the organelle in a similar ... More
Novel Fusogenic Liposomes for Fluorescent Cell Labeling and Membrane Modification.
Authors:Csisza´r A, Hersch N, Dieluweit S, Biehl R, Merkel R, Hoffmann B,
Journal:Bioconjug Chem
PubMed ID:20184308
'Efficient delivery of biomolecules into membranes of living cells as well as cell surface modifications are major biotechnological challenges. Here, novel liposome systems based on neutral and cationic lipids in combination with lipids modified by aromatic groups are introduced for such applications. The fusion efficiency of these liposome systems was ... More
Group V phospholipase A2 induces leukotriene biosynthesis in human neutrophils through the activation of group IVA phospholipase A2.
Authors:Kim YJ, Kim KP, Han SK, Munoz NM, Zhu X, Sano H, Leff AR, Cho W
Journal:J Biol Chem
PubMed ID:12124392
'We reported previously that exogenously added human group V phospholipase A(2) (hVPLA(2)) could elicit leukotriene B(4) (LTB(4)) biosynthesis in human neutrophils (Han, S. K., Kim, K. P., Koduri, R., Bittova, L., Munoz, N. M., Leff, A. R., Wilton, D. C., Gelb, M. H., and Cho, W. (1999) J. Biol. Chem. ... More
Determination of the depth of BODIPY probes in model membranes by parallax analysis of fluorescence quenching.
Authors:Kaiser RD, London E
Journal:Biochim Biophys Acta
PubMed ID:9767081
'The location of a series of lipophilic and lipid-attached BODIPY (4, 4-difluoro-4-bora-3a,4a-diaza-s-indacene) membrane probes was analyzed by the quenching of BODIPY fluorescence by a series of nitroxide-labeled lipids in which the depth of the nitroxide group is varied. When attached to the polar headgroup of PE the BODIPY remained near ... More
The antimicrobial peptide trichogin and its interaction with phospholipid membranes.
Authors:Epand RF, Epand RM, Monaco V, Stoia S, Formaggio F, Crisma M, Toniolo C
Journal:Eur J Biochem
PubMed ID:10583397
'The interaction of the antimicrobial peptide trichogin GA IV with phospholipid bilayers has been studied. A series of analogs of trichogin was synthesized in which the nitroxide spin label, 4-amino-4-carboxy-2,2,6,6-tetramethylpiperidino-1-oxyl (TOAC), replaced one of the three alpha-aminoisobutyric acid (Aib) residues in the sequence. These modified peptides were used to assess ... More