BODIPY™ 581/591 C11 (Lipid Peroxidation Sensor) - FAQs
BODIPY™ 581/591 C11 (Lipid Peroxidation Sensor) - FAQs
View additional product information for BODIPY™ 581/591 C11 (Lipid Peroxidation Sensor) - FAQs (D3861)
1 product FAQs found
What solvent should be used to reconstitute/dissolve BODIPY 581/591 C11 (Lipid Peroxidation Sensor) (Cat. No. D3861)?
For BODIPY 581/591 C11 (Cat No. D3861), we recommend that you dissolve this in dimethyl sulfoxide (DMSO). You can prepare a stock solution in any concentration between 1 and 5 mg/mL. Please note that the BODIPY/DMSO stock solution must be stored at -20 degrees C, desiccated and protected from light.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.