Owl™ D4 Horizontal Electrophoresis System Replacement Parts
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
Have Questions?
Change view
Catalog Number
Type
D4-CST
Caster
D4-GK
Gasket
D4-UVT
Gel Tray
3 Options
Catalog number D4-CST
Price (USD)
357.30
Online Exclusive
397.00
Save 39.70 (10%)
Each
Type:
Caster
Price (USD)
357.30
Online Exclusive
397.00
Save 39.70 (10%)
Each
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
Specifications
Length (Metric)17 cm
Width (Metric)16 cm
For Use With (Equipment)Owl™ D4 Electrophoresis System