Product Image
Thermo Scientific™
Harvey™ DI+ Cartridge System with replacement cartridge
Remove calcium, magnesium and other ionic impurities that can harm steam sterilizers. This system is quick to set up, easy to use, and requires only simple maintance.
Have Questions?
Número de catálogo D502118
Precio (EUR)
-
Remove calcium, magnesium and other ionic impurities that can harm steam sterilizers with Thermo Scientific™ Harvey DI Cartridge System. It is quick to set up, easy to use and requires only simple maintance. This system mounts to the wall for easy installation and eliminates the need to store containers of distilled water.
  • Quick-fitting connectors hook directly to faucet—no power required
  • Nuclear-grade resin is enclosed in a virgin polypropylene canister to purify tap water
  • Produces up to 4L of pure water per minute
  • 23 cents per gal. cost based on average tap water (171ppm)
  • Simple maintenance: quarter-turn quick-release canister is easy to replace
  • Pura-lite notifies when it is time for a new cartridge
Especificaciones
DescripciónCartucho de repuesto Harvey DI+
TipoDI+ Cartridge System
Caudal4 l/min
Unit SizeEach