Barnstead™ Easypure™ Cartridges and Filters
Actual product may vary
Thermo Scientific™
Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeFilterSizedimResinType
D502124Easypure II UV, Easypure II UV/UFCartridge Pack, Deionized Feed
D502125Easypure II UV, Easypure II UV/UFCartridge Pack, RO or Distilled Feed
D502126Easypure II, Easypure UFCartridge Pack, Deionized Feed
D502127Easypure II, Easypure UFCartridge Pack, RO or Distilled Feed
Catalog number D502124
Price (USD)
727.20
Online Exclusive
808.00
Save 80.80 (10%)
Each
For Use With (Equipment):
Easypure II UV, Easypure II UV/UF
Type:
Cartridge Pack, Deionized Feed
Price (USD)
727.20
Online Exclusive
808.00
Save 80.80 (10%)
Each
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
  • Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
  • Type 1 filter
D502124:
  • Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
D502127:
  • Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
  • Type 1 filter
D502125:
  • Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
Specifications
For Use With (Equipment)Easypure II UV, Easypure II UV/UF
TypeCartridge Pack, Deionized Feed
Feedwater SourceDeionized
Unit SizeEach