Barnstead™ Easypure™ II
El producto real puede variar
Thermo Scientific™

Barnstead™ Easypure™ II

This product has been discontinued. For additional information or alternate products please contact your sales representative.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripción
D7031Discontinuado; el producto de sustitución es 50132367
D7035Discontinuado; el producto de sustitución es 50132374
D7381Discontinuado; el producto de sustitución es 50132366
D7401Discontinuado; el producto de sustitución es 50132373
D7411Discontinuado; el producto de sustitución es 50132368
D8611Discontinuado; el producto de sustitución es 50132370
Número de catálogo D7031
Precio (MXN)
-
Descripción:
Discontinuado; el producto de sustitución es 50132367

This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is Thermo Scientific™ Barnstead™ MicroPure™.

Features:
  • Each system produces ultrapure water that exceeds the quantitative specifications of Type 1 water as described in ASTM™ D1193, ISO 3696, and CLSI-CLRW.
  • Three cartridges in series provide increased capacity over similar systems.
  • A 0.2μm final filter at the point of use provides water free of particles and bacteria.
  • Quiet pump provides optimal flowrates and silent operation.
  • Standby mode recirculates purified water during inactivity to ensure purity and allows for on-demand use.
  • All components are constructed of inert materials, ensuring maximum ionic and organic purity.
  • Supplied wall bracket allows increased flexibility when choosing a location.
  • Integral low water and low pressure pump protectors protect the pump in the event feedwater is interrupted to the system.

Easypure II:

  • Base unit
  • For buffer prep, titration, solutions, AA
  • TOC 5 to 10ppb

Easypure II UV:

  • Ultralow organics
  • For HPLC, TOC, GC/MS
  • TOC 1 to 5ppb

Easypure II UFI:

  • Eliminates pyrogens
  • For cell and tissue culture, monoclonal antibody production, electrophoresis
  • TOC 5 to 10ppb

Easypure II UV:

  • Lowest levels of organics, pyrogens and nucleases
  • For PCR 2-D electrophoresis, cell culture, blotting
  • TOC 1 to 5ppb

Ordering Information:

Optional accessories include Remote Dispenser (0903515) and replacement consumables (UV lamps, ultrafilters and cleaning syringe); order separately. Contact your Sales Representative for details.

Includes:

Wall mounting bracket, cleaning syringe, UV lamp (Easypure II UV and UV/UF only), ultrafilter (Easypure II UF and UV/UF only).

Requires:

  • Cartridges and filters

Recommended for:

  • HPLC
  • ICP
  • GC/MS
  • Cell and tissue culture
  • PCR

Note:

  • Feedwater must be pre-treated by reverse osmosis, distillation or deionization
Especificaciones
Certificaciones/conformidadCE, CSA
Conductividad0.055 µS/cm
DescripciónDiscontinuado; el producto de sustitución es 50132367
Requisitos eléctricos100/240 VCA, 50/60 Hz
Presión del agua de suministro100 psig (6,9 bar)
Temperatura del agua de suministro (métrico)4°C a 40°C
Frecuencia50/60 Hz
ModeloRF model
Partículas > 0,2 µM/ml<1 μM/mL
Tipo de productoSistema de purificación de agua
Tensión100/240 VCA
Contenido bacteriano<1 CFU/mL
Monitorización del agua de suministroNo
Fuente del agua de suministroPretratado
CaudalHasta 1 l/min
Tipo de agua productoTipo 1
Resistividad18,2 Mohm/cm a 25 °C
TOC Monitor with UV Intensity MonitoringNo
Carbono orgánico total (COT)0 to 10 ppb
Unit Size-