Barnstead™ Easypure™ II
Actual product may vary
Thermo Scientific™

Barnstead™ Easypure™ II

This product has been discontinued. For additional information or alternate products please contact your sales representative.
Have Questions?
變更視圖buttonViewtableView
產品號碼Description
D8611Discontinued; replacement product is 50132370
D7031Discontinued; replacement product is 50132367
D7035Discontinued; replacement product is 50132374
D7381Discontinued; replacement product is 50132366
D7401Discontinued; replacement product is 50132373
D7411Discontinued; replacement product is 50132368
產品號碼 D8611
價格 (HKD)
-
Description:
Discontinued; replacement product is 50132370

This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is Thermo Scientific™ Barnstead™ MicroPure™.

Features:
  • Each system produces ultrapure water that exceeds the quantitative specifications of Type 1 water as described in ASTM™ D1193, ISO 3696, and CLSI-CLRW.
  • Three cartridges in series provide increased capacity over similar systems.
  • A 0.2μm final filter at the point of use provides water free of particles and bacteria.
  • Quiet pump provides optimal flowrates and silent operation.
  • Standby mode recirculates purified water during inactivity to ensure purity and allows for on-demand use.
  • All components are constructed of inert materials, ensuring maximum ionic and organic purity.
  • Supplied wall bracket allows increased flexibility when choosing a location.
  • Integral low water and low pressure pump protectors protect the pump in the event feedwater is interrupted to the system.

Easypure II:

  • Base unit
  • For buffer prep, titration, solutions, AA
  • TOC 5 to 10ppb

Easypure II UV:

  • Ultralow organics
  • For HPLC, TOC, GC/MS
  • TOC 1 to 5ppb

Easypure II UFI:

  • Eliminates pyrogens
  • For cell and tissue culture, monoclonal antibody production, electrophoresis
  • TOC 5 to 10ppb

Easypure II UV:

  • Lowest levels of organics, pyrogens and nucleases
  • For PCR 2-D electrophoresis, cell culture, blotting
  • TOC 1 to 5ppb

Ordering Information:

Optional accessories include Remote Dispenser (0903515) and replacement consumables (UV lamps, ultrafilters and cleaning syringe); order separately. Contact your Sales Representative for details.

Includes:

Wall mounting bracket, cleaning syringe, UV lamp (Easypure II UV and UV/UF only), ultrafilter (Easypure II UF and UV/UF only).

Requires:

  • Cartridges and filters

Recommended for:

  • HPLC
  • ICP
  • GC/MS
  • Cell and tissue culture
  • PCR

Note:

  • Feedwater must be pre-treated by reverse osmosis, distillation or deionization
規格
Certifications/ComplianceCE, CSA
Conductivity0.055 µS/cm
DescriptionDiscontinued; replacement product is 50132370
Electrical Requirements100/240 V 50/60 Hz
Endotoxin Concentration0.001 EU/mL
Feedwater Pressure100 psig (6.9 bar)
Feedwater Temperature (Metric)4°C to 40°C
Frequency50/60 Hz
ModelUV/UF model
Particles >0.2µM/mL<1 μM/mL
Product TypeWater Purification System
Voltage100/240 V
Bacterial Content<1 CFU/mL
Feed Water MonitoringNo
Feedwater SourcePretreated
Flow RateUp to 1 L/min.
Product Water TypeType 1
Resistivity18.2 Mohm.cm at 25°C
TOC Monitor with UV Intensity MonitoringNo
Total Organic Carbon1 to 5 ppb
Unit Size-