Reverse Osmosis Membranes
Actual product may vary
Thermo Scientific™
Reverse Osmosis Membranes
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
D9006Reverse Osmosis MembraneROpure Infinity Systems
22.0108Reverse Osmosis MembraneMediTower 140
22.0087Reverse Osmosis MembraneLabTower EDI 30, LabTower RO 40/60, LabTower TII 40/60, Pacific RO 40, Pacific TII 40
09.2003Reverse Osmosis Membrane (pretreated), 3 L/hr.Smart2Pure
09.2006Reverse Osmosis Membrane (pretreated), 6 L/hr.Smart2Pure
22.0046Reverse Osmosis MembraneLabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
Catalog number D9006
Price (MXN)
-
Type:
Reverse Osmosis Membrane
For Use With (Equipment):
ROpure Infinity Systems
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
  • Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
  • Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
  • LabTower™ EDI 15 requires two of these RO membranes
  • LabTower™ RO 20 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • LabTower™ TII 20 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • Pacific™ RO 3, 7, 12 require one of these RO membranes
  • Pacific RO 20 requires two of these RO membranes
  • Pacific™ TII 3, 7, 12 require one of these RO membranes
  • Pacific TII 20 requires two of these RO membranes
22.0087:
  • LabTower™ EDI 30 requires two of these RO membranes
  • LabTower™ RO 40 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • LabTower TII 40 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • Pacific RO 40 requires two of these RO membranes
  • Pacific TII 40 requires two of these RO membranes
Specifications
For Use With (Equipment)ROpure Infinity Systems
TypeReverse Osmosis Membrane
Unit SizeEach