Owl™ Casting Dams
Owl™ Casting Dams
Thermo Scientific™
Owl™ Casting Dams
Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (Metric)
DAM-1313 cm
DAM-2020 cm
DAM-2323 cm
Catalog number DAM-13
Price (MXN)
-
Depth (Metric):
13 cm
Block off a portion of the gel tray and allow shorter gels or gradient gels to be cast and run with the high-quality aluminum Thermo Scientific™ Owl™ Casting Dams.
  • Seal agarose on contact, eliminating the need for tape
  • Does not attach to gel tray, enabling gels of any length to be poured
  • Easily handled and maneuvered using knob or finger depressions
  • Dam blocks off a portion of UVT gel tray
Specifications
Depth (Metric)13 cm
DescriptionCasting Dam, 13cm (L)
For Use With (Equipment)Owl System
Product LineOwl
TypeGel Casting Dam
Unit SizeEach