Thermo Scientific™
Dissecting Board
 Related applications:
Anatomical Pathology
No swelling, warping or cracking when you use Thermo Scientific™ Richard-Allan Scientific™ Dissecting Boards, crafted of high-density polyethylene.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Features:
  • Dimensional scales permit quick sizing and serve as part of permanent record
  • Rubber feet stabilize working surface and help prevent board from slipping on workbench
  • White background contrasts with specimens for excellent photographic detail
  • Boards have a spill-containment border
  • Styrene-capped pins secure specimens and are available separately
Dimensions:
  • Large board measures 13 x 11 x 0.63 in. (33 x 28 x 1.6cm)
  • Small board measures 23 x 16 x 0.38 in. (58.4 x 40.6 x 0.97cm)
Documents
Manuals & protocols